Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL20RB Mouse Monoclonal Antibody [clone: 01]

Supplier: Sino Biological

80187-MM01-100 80187-MM01-200 80187-MM01-50
103661-288EA 160.46 USD InStock
103661-288 103661-290 103596-956
Anti-IL20RB Mouse Monoclonal Antibody [clone: 01]
Antibodies
Anti-IL20RB Antibody

Type: Primary
Antigen: IL20RB
Clonality: Monoclonal
Clone: Clone: 01
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty