Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LTBR Rabbit Polyclonal Antibody

Supplier: Sino Biological

80178-RP01-100 80178-RP01-400 80178-RP01-200
103662-586EA 106.62 USD InStock
103662-586 103662-590 103662-588
Anti-LTBR Rabbit Polyclonal Antibody
Antibodies
Anti-LTBR Antibody

Type: Primary
Antigen: LTBR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty