Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IFNA1 Mouse Monoclonal Antibody [clone: 02]

Supplier: Sino Biological

80174-MM02-50 80174-MM02-100 80174-MM02-200
103596-910EA 106.62 USD InStock
103596-910 103661-280 103661-282
Anti-IFNA1 Mouse Monoclonal Antibody [clone: 02]
Antibodies
Anti-Interferon alpha 1 Antibo

Type: Primary
Antigen: IFNA1
Clonality: Monoclonal
Clone: Clone: 02
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty