Anti-TNFSF9 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80166-T16-100 80166-T16-200 80166-T16-50
103596-902EA 270.87 USD
103596-902 103596-904 103637-586
Anti-TNFSF9 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat 4-1BBL/CD137L (Catalog#80166-R07H; Q80WE6; Pro106-Gly308). 4-1BBL/CD137L specific IgG was purified by Rat 4-1BBL/CD137L affinity chromatography.

Type: Primary
Antigen: 4-1BBL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR