Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-4-1BBL Rabbit Polyclonal Antibody

Supplier: Sino Biological

80166-RP01-400 80166-RP01-100 80166-RP01-200
103661-278EA 214.31 USD InStock
103661-278 103661-276 103661-274
Anti-4-1BBL Rabbit Polyclonal Antibody
Antibodies
Anti-4-1BBL Antibody

Type: Primary
Antigen: 4-1BBL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty