Anti-RANK Rabbit Polyclonal Antibody

Supplier: Sino Biological

80160-T16-200 80160-T16-100 80160-T16-50
103596-900EA 214.31 USD
103596-900 103596-898 103637-584
Anti-RANK Rabbit Polyclonal Antibody
Antibodies
Anti-RANK Antibody

Type: Primary
Antigen: RANK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR