Anti-RANK Rabbit Monoclonal Antibody [clone: 044]

Supplier: Sino Biological

80160-R044-50 80160-R044-100
103596-896EA 128.15 USD
103596-896 103662-576
Anti-RANK Rabbit Monoclonal Antibody [clone: 044]
Antibodies
Anti-RANK Antibody

Type: Primary
Antigen: RANK
Clonality: Monoclonal
Clone: Clone: 044
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR