Anti-CD40 Mouse Monoclonal Antibody [clone: 11]

Supplier: Sino Biological
80151-MM11-50 80151-MM11-200 80151-MM11-100
103596-890EA 142.96 USD
103596-890 103661-264 103661-266
Anti-CD40 Mouse Monoclonal Antibody [clone: 11]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat CD40 / TNFRSF5 (rR CD40 / TNFRSF5; Catalog#80151-R08H; NP_599187.1; Met1-Arg193). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: Anti-CD40 Antibody
Clonality: Monoclonal
Clone: 11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR