Anti-Anti-CD40 Antibody Mouse Monoclonal Antibody [clone: 01]

Supplier: Sino Biological

80151-MM01-50 80151-MM01-100 80151-MM01-200
103596-888EA 106.62 USD
103596-888 103661-260 103661-262
Anti-Anti-CD40 Antibody Mouse Monoclonal Antibody [clone: 01]
Antibodies
Anti-CD40 Antibody

Type: Primary
Antigen: Anti-CD40 Antibody
Clonality: Monoclonal
Clone: Clone: 01
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR