Anti-CXCL16 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80149-T16-200 80149-T16-50 80149-T16-100
103596-882EA 214.31 USD
103596-882 103637-578 103596-880
Anti-CXCL16 Rabbit Polyclonal Antibody
Antibodies
Anti-CXCL16 Antibody

Type: Primary
Antigen: CXCL16
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR