Anti-EFNB2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80107-T16-100 80107-T16-50 80107-T16-200
103596-858EA 270.87 USD
103596-858 103589-734 103596-860
Anti-EFNB2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat Ephrin-B2 (Catalog#80107-R08H; NP_001100798.1; Met1-Ala229). Ephrin-B2 specific IgG was purified by Rat Ephrin-B2 affinity chromatography.

Type: Primary
Antigen: EPHB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR