Anti-EFNB1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80106-T52-50 80106-T52-200 80106-T52-100
103596-854EA 270.87 USD
103596-854 103596-856 103589-732
Anti-EFNB1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat Ephrin-B1/EFNB1 (Catalog#80106-R08H; P52796; Met1-Thr229). Ephrin-B1/EFNB1 specific IgG was purified by Rat Ephrin-B1/EFNB1 affinity chromatography.

Type: Primary
Antigen: EPHB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR