Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FLT1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80101-RP01-200 80101-RP01-100 80101-RP01-400
103661-226EA 160.46 USD InStock
103661-226 103661-228 103661-230
Anti-FLT1 Rabbit Polyclonal Antibody
Antibodies
Anti-FLT1 Antibody

Type: Primary
Antigen: FLT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty