Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EGFR Rabbit Polyclonal Antibody

Supplier: Sino Biological

80100-T52-50 80100-T52-100 80100-T52-200
103589-728EA 106.62 USD InStock
103589-728 103596-844 103596-846
Anti-EGFR Rabbit Polyclonal Antibody
Antibodies
Anti-EGFR Antibody

Type: Primary
Antigen: EGFR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty