Anti-PGC Rabbit Polyclonal Antibody

Supplier: Sino Biological
80097-RP01-400 80097-RP01-100 80097-RP01-200
103661-218EA 562.97 USD
103661-218 103662-574 103662-572
Anti-PGC Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat Pepsinogen C/PGC (Catalog#80097-R08H; NP_579818.1; Met1-Val392). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: PGC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR