Anti-PGC Rabbit Monoclonal Antibody [clone: 008]

Supplier: Sino Biological

80097-R008-100 80097-R008-50
103662-570EA 214.31 USD
103662-570 103596-838
Anti-PGC Rabbit Monoclonal Antibody [clone: 008]
Antibodies
Anti-Pepsinogen II Antibody

Type: Primary
Antigen: PGC
Clonality: Monoclonal
Clone: Clone: 008
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR