Anti-FGFR4 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80093-RP02-50 80093-RP02-100 80093-RP02-200
103596-836EA 142.96 USD
103596-836 103661-042 103661-216
Anti-FGFR4 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat FGFR4/FGF Receptor 4 (rR FGFR4/FGF Receptor 4; Catalog#80093-R08H; Q498D6; Met1-Asp367). FGFR4/FGF Receptor 4 specific IgG was purified by Rat FGFR4/FGF Receptor 4 affinity chromatography.

Type: Primary
Antigen: FGFR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR