Anti-UCHL1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80089-RP02-100 80089-RP02-50 80089-RP02-200
103596-832EA 99 USD
103596-832 103661-210 103661-212
Anti-UCHL1 Rabbit Polyclonal Antibody
Antibodies
Anti-UCHL1 Antibody

Type: Primary
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR