Anti-UCHL1 Rabbit Monoclonal Antibody [clone: 012]

Supplier: Sino Biological

80089-R012-100 80089-R012-50
103661-202EA 214.31 USD
103661-202 103596-830
Anti-UCHL1 Rabbit Monoclonal Antibody [clone: 012]
Antibodies
Anti-UCHL1 Antibody

Type: Primary
Antigen: UCHL1
Clonality: Monoclonal
Clone: Clone: 012
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR