Anti-UCHL3 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80087-RP02-200 80087-RP02-100 80087-RP02-50
103661-200EA 214.31 USD
103661-200 103661-198 103596-828
Anti-UCHL3 Rabbit Polyclonal Antibody
Antibodies
Anti-UCHL3 Antibody

Type: Primary
Antigen: UCHL3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR