Anti-UCHL3 Rabbit Monoclonal Antibody [clone: 007]

Supplier: Sino Biological
80087-R007-100 80087-R007-50
103661-194EA 271.4 USD
103661-194 103596-824
Anti-UCHL3 Rabbit Monoclonal Antibody [clone: 007]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat UCHL3 (rR UCHL3; Catalog#80087-R07E; Q91Y78; Glu2-Ala230).

Type: Primary
Antigen: UCHL3
Clonality: Monoclonal
Clone: 007
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR