Anti-CD4 Mouse Monoclonal Antibody [clone: 05]

Supplier: Sino Biological
80086-MM05-100 80086-MM05-50 80086-MM05-200
103661-190EA 270.87 USD
103661-190 103596-822 103661-192
Anti-CD4 Mouse Monoclonal Antibody [clone: 05]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat CD4 (rR CD4; Catalog#80086-R08H; NP_036837.1; Met1-Thr394). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: Anti-CD4 Antibody
Clonality: Monoclonal
Clone: 05
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR