Anti-WFDC2 Rabbit Monoclonal Antibody [clone: 004]

Supplier: Sino Biological
80084-R004-100 80084-R004-50
103596-816EA 150.18 USD
103596-816 103661-188
Anti-WFDC2 Rabbit Monoclonal Antibody [clone: 004]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat HE4 / WFDC2 / WAP5 (rR HE4 / WFDC2 / WAP5; CAD56203.1; aa1-120).

Type: Primary
Antigen: HE4
Clonality: Monoclonal
Clone: 004
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR