Anti-IL17RA Rabbit Polyclonal Antibody

Supplier: Sino Biological

80083-T16-50 80083-T16-100 80083-T16-200
103589-720EA 106.62 USD
103589-720 103596-812 103596-814
Anti-IL17RA Rabbit Polyclonal Antibody
Antibodies
Anti-IL7R alpha Antibody

Type: Primary
Antigen: IL17RA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR