Anti-IL7R Rabbit Polyclonal Antibody

Supplier: Sino Biological
80083-RP01-100 80083-RP01-400 80083-RP01-200
103661-184EA 270.2 USD
103661-184 103661-186 103661-182
Anti-IL7R Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat CD127 / IL-7RA (Catalog#80083-R08H; NP_001099888.1; Met1-Asp239). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: IL17RA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR