Anti-ERBB2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80079-T52-50 80079-T52-200 80079-T52-100
103589-718EA 142.96 USD
103589-718 103596-810 103596-808
Anti-ERBB2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat HER2/ErbB2 (rR HER2/ErbB2; Catalog#80079-R08H; AAH61863.1; Met4-Thr656). HER2/ErbB2 specific IgG was purified by Rat HER2/ErbB2 affinity chromatography.

Type: Primary
Antigen: Her2 / ERBB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR