Anti-Her2 / ERBB2 Rabbit Monoclonal Antibody [clone: 187]

Supplier: Sino Biological

80079-R187-50 80079-R187-100
103596-802EA 128.15 USD
103596-802 103661-174
Anti-Her2 / ERBB2 Rabbit Monoclonal Antibody [clone: 187]
Antibodies
Anti-Her2 / ERBB2 Antibody

Type: Primary
Antigen: Her2 / ERBB2
Clonality: Monoclonal
Clone: Clone: 187
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR