Anti-IL2 Mouse Monoclonal Antibody [clone: 10]

Supplier: Sino Biological
80075-MM10-50 80075-MM10-100 80075-MM10-200
103661-160EA 352.03 USD
103661-160 103661-158 103596-782
Anti-IL2 Mouse Monoclonal Antibody [clone: 10]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat Interleukin-2 / IL-2 (rR Interleukin-2 / IL-2; Catalog#80075-RNAE; P17108; Ala21-Gln155). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: IL2
Clonality: Monoclonal
Clone: 10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR