Anti-PRLR Rabbit Polyclonal Antibody

Supplier: Sino Biological
80062-T16-100 80062-T16-50 80062-T16-200
103596-778EA 270.87 USD
103596-778 103589-708 103596-780
Anti-PRLR Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat Prolactin Receptor (Catalog#80062-R08H; P05710-1; Met1-Asp229). Prolactin Receptor specific IgG was purified by Rat Prolactin Receptor affinity chromatography.

Type: Primary
Antigen: PRLR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR