Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PRLR Rabbit Polyclonal Antibody

Supplier: Sino Biological

80062-RP01-200 80062-RP01-100 80062-RP01-400
103661-154EA 160.46 USD InStock
103661-154 103661-152 103661-156
Anti-PRLR Rabbit Polyclonal Antibody
Antibodies
Anti-Prolactin Receptor Antibo

Type: Primary
Antigen: PRLR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty