Anti-LCN2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80058-RP02-100 80058-RP02-50 80058-RP02-200
103661-144EA 270.87 USD
103661-144 103661-146 103596-772
Anti-LCN2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat LCN2 / NGAL (rR LCN2 / NGAL; Catalog#80058-R08B; P30152; Met1-Asn198). LCN2 / NGAL specific IgG was purified by Rat LCN2 / NGAL affinity chromatography.

Type: Primary
Antigen: LCN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR