Anti-LCN2 Mouse Monoclonal Antibody [clone: 04]

Supplier: Sino Biological

80058-MM04-50 80058-MM04-200 80058-MM04-100
103596-768EA 106.62 USD
103596-768 103661-134 103661-132
Anti-LCN2 Mouse Monoclonal Antibody [clone: 04]
Antibodies
Anti-Lipocalin-2 Antibody

Type: Primary
Antigen: LCN2
Clonality: Monoclonal
Clone: Clone: 04
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR