Anti-EPO Rabbit Polyclonal Antibody

Supplier: Sino Biological

80055-T16-200 80055-T16-50 80055-T16-100
103596-766EA 214.31 USD
103596-766 103589-706 103596-764
Anti-EPO Rabbit Polyclonal Antibody
Antibodies
Anti-Erythropoietin Antibody

Type: Primary
Antigen: EPO
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR