Anti-MMP9 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80049-T24-50 80049-T24-100 80049-T24-200
103589-704EA 142.96 USD
103589-704 103596-758 103596-760
Anti-MMP9 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat MMP-9/MMP9 (rR MMP-9/MMP9; Catalog#80049-R08H; EDL96479.1; Met1-Pro708). MMP-9/MMP9 specific IgG was purified by Rat MMP-9/MMP9 affinity chromatography.

Type: Primary
Antigen: MMP-9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR