Anti-MMP9 Rabbit Monoclonal Antibody [clone: 213]

Supplier: Sino Biological
80049-R213-50 80049-R213-100
103661-032EA 271.4 USD
103661-032 103596-754
Anti-MMP9 Rabbit Monoclonal Antibody [clone: 213]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat MMP9/MMP-9/CLG4B (rR MMP9/MMP-9/CLG4B; Catalog#80049-R08H; EDL96479.1; Met1-Pro708).

Type: Primary
Antigen: MMP-9
Clonality: Monoclonal
Clone: 213
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR