Anti-MMP-9 Mouse Monoclonal Antibody [clone: 01]

Supplier: Sino Biological

80049-MM01-1 80049-MM01-200 80049-MM01-50 80049-MM01-100
103589-702EA 958.46 USD
103589-702 103661-026 103596-750 103661-120
Anti-MMP-9 Mouse Monoclonal Antibody [clone: 01]
Antibodies
Anti-MMP-9 Antibody

Type: Primary
Antigen: MMP-9
Clonality: Monoclonal
Clone: Clone: 01
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR