Anti-NXPH1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80047-RP01-200 80047-RP01-400 80047-RP01-100
103661-118EA 562.97 USD
103661-118 103661-106 103661-116
Anti-NXPH1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat Neurexophilin-1/NXPH1 (Catalog#80047-R08H; Q63366-1; Met1-Gly271). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: NXPH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR