Anti-NXPH1 Rabbit Monoclonal Antibody [clone: 001]

Supplier: Sino Biological
80047-R001-50 80047-R001-100
103596-742EA 150.18 USD
103596-742 103661-102
Anti-NXPH1 Rabbit Monoclonal Antibody [clone: 001]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat Neurexophilin-1 / NXPH1 (rR Neurexophilin-1 / NXPH1; Catalog#80047-R08H; Q63366-1; Met1-Gly271).

Type: Primary
Antigen: NXPH1
Clonality: Monoclonal
Clone: 001
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR