Anti-TNFA Rabbit Polyclonal Antibody

Supplier: Sino Biological

80045-RP01-400 80045-RP01-100 80045-RP01-200
103661-096EA 214.31 USD
103661-096 103661-094 103661-092
Anti-TNFA Rabbit Polyclonal Antibody
Antibodies
Anti-TNF-alpha Antibody

Type: Primary
Antigen: TNFA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR