Anti-MIP-1 alpha Rabbit Polyclonal Antibody

Supplier: Sino Biological

80044-T32-100 80044-T32-50 80044-T32-200
103596-728EA 160.46 USD
103596-728 103589-694 103596-730
Anti-MIP-1 alpha Rabbit Polyclonal Antibody
Antibodies
Anti-MIP-1 alpha Antibody

Type: Primary
Antigen: MIP-1 alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR