Anti-MIP-1 alpha Rabbit Monoclonal Antibody [clone: 013]

Supplier: Sino Biological

80044-R013-100 80044-R013-50
103661-090EA 214.31 USD
103661-090 103596-726
Anti-MIP-1 alpha Rabbit Monoclonal Antibody [clone: 013]
Antibodies
Anti-MIP-1 alpha Antibody

Type: Primary
Antigen: MIP-1 alpha
Clonality: Monoclonal
Clone: Clone: 013
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR