Anti-FAS Rabbit Polyclonal Antibody

Supplier: Sino Biological
80043-T16-200 80043-T16-50 80043-T16-100
103596-722EA 270.87 USD
103596-722 103589-692 103596-724
Anti-FAS Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat CD95 / APO-1 / TNFRSF6 (Catalog#80043-R08H; NP_631933.2; Met1-Lys170). CD95 / APO-1 / TNFRSF6 specific IgG was purified by Rat CD95 / APO-1 / TNFRSF6 affinity chromatography.

Type: Primary
Antigen: TNFRSF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR