Anti-FAS Rabbit Monoclonal Antibody [clone: 105]

Supplier: Sino Biological
80043-R105-100 80043-R105-50
103661-082EA 271.4 USD
103661-082 103596-720
Anti-FAS Rabbit Monoclonal Antibody [clone: 105]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat FAS / TNFRSF6 / CD95 (rR FAS / TNFRSF6 / CD95; Catalog#80043-R08H; NP_631933.2; Met1-Lys170).

Type: Primary
Antigen: TNFRSF6
Clonality: Monoclonal
Clone: 105
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR