Anti-FAS Rabbit Monoclonal Antibody [clone: 028]

Supplier: Sino Biological
80043-R028-50 80043-R028-100
103596-718EA 150.18 USD
103596-718 103661-024
Anti-FAS Rabbit Monoclonal Antibody [clone: 028]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD95 / APO-1 / TNFRSF6 / FAS (rR CD95 / APO-1 / TNFRSF6 / FAS; Catalog#80043-R08H; NP_631933.2; Met1-Lys170).

Type: Primary
Antigen: TNFRSF6
Clonality: Monoclonal
Clone: 028
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR