Anti-FAS Rabbit Monoclonal Antibody [clone: 018]

Supplier: Sino Biological
80043-R018-100 80043-R018-50
103596-716EA 150.18 USD
103596-716 103661-022
Anti-FAS Rabbit Monoclonal Antibody [clone: 018]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat FAS/TNFRSF6/CD95 (rR FAS/TNFRSF6/CD95; Catalog#80043-R08H; NP_631933.2; Met1-Lys170).

Type: Primary
Antigen: TNFRSF6
Clonality: Monoclonal
Clone: 018
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR