Anti-CXCL2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80042-RP01-100 80042-RP01-400 80042-RP01-200
103661-020EA 562.97 USD
103661-020 103661-016 103661-018
Anti-CXCL2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat CXCL2/MIP-2 (rR CXCL2/MIP-2; Catalog#80042-RNCE; NP_446099.1; Ser32-Asn100). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CXCL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR