Anti-CXCL2 Rabbit Monoclonal Antibody [clone: 126]

Supplier: Sino Biological
80042-R126-50 80042-R126-100
103596-710EA 150.18 USD
103596-710 103661-014
Anti-CXCL2 Rabbit Monoclonal Antibody [clone: 126]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CXCL2/MIP-2 (rR CXCL2/MIP-2; Catalog#80042-RNCE; NP_446099.1; Ser32-Asn100).

Type: Primary
Antigen: CXCL2
Clonality: Monoclonal
Clone: 126
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR