Anti-CRP Rabbit Polyclonal Antibody

Supplier: Sino Biological

80041-RP02-100 80041-RP02-50 80041-RP02-200
103661-010EA 160.46 USD
103661-010 103596-708 103661-012
Anti-CRP Rabbit Polyclonal Antibody
Antibodies
Anti-C-Reactive Protein Antibo

Type: Primary
Antigen: CRP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR