Anti-CRP Rabbit Polyclonal Antibody

Supplier: Sino Biological
80041-RP01-400 80041-RP01-200 80041-RP01-100
103661-076EA 270.2 USD
103661-076 103661-080 103661-078
Anti-CRP Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat CRP / C-Reactive Protein (Catalog#80041-R08H; NP_058792.1; Met1-Ser230). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CRP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR