Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CSTC Rabbit Polyclonal Antibody

Supplier: Sino Biological

80040-RP02-100 80040-RP02-50 80040-RP02-200
103661-072EA 160.46 USD InStock
103661-072 103596-702 103661-074
Anti-CSTC Rabbit Polyclonal Antibody
Antibodies
Anti-Cystatin C Antibody

Type: Primary
Antigen: CSTC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty